Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, dNTPs

VWR®, dNTPs

dNTP mix, 10 mM of each dA, dC, dG, and dT, 2×500 μl, 1.000 µl
Catalog # 733-1363
Supplier:   VWR Chemicals
VWR®, dNTPs
Catalog # 733-1363
Supplier:  VWR Chemicals
CAS Number:  
VWR®, dNTPs
VWR®, dNTPs

dNTP mix, 10 mM of each dA, dC, dG, and dT, 2×500 μl, 1.000 µl

CAS Number:  
Price
80,60 €
Log in for availability
1 * 1.000 µl
Product Details & Documents

Ready to use molecular biology grade dNTP mixes and dNTP sets. The dNTP mix is designed to save hands-on time for researchers, and reduce the possibility of contamination by reducing pipetting. dNTP solutions are also available in sets of four individual dNTPs, each 100 mm. Both are convenient for use in DNA polymerisation reactions, DNA labelling and sequencing processes.

  • Available as pre-mixed 10 or 25 mM solutions, or as sets of individual 100 mM dNTP solutions
  • Both pre-mixed and sets have been functionally tested in PCR
  • Purity >99% by HPLC
  • Supplied in solution at pH 7,3 to 7,5
Specifications

Specifications

  • Pk:
    1.000 µl
  • Description:
    dNTP mix, 10 mM of each dA, dC, dG, and dT, 2×500 μl

Specifications