Orders LinkContactUsLinkComponent
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining rack, only, for 10 to 20 slides, - mm
Catalog # 631-2504
Supplier:   DWK Life Sciences
Staining dishes, WHEATON®
Catalog # 631-2504
Supplier:  DWK Life Sciences
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining rack, only, for 10 to 20 slides, - mm
Price
167,00 €
Log in for availability
1 * 3 ST
Product Details & Documents
Soda-lime glass and stainless steel.
  • Removable glass slide rack has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
Standard for manual staining procedures. The rack holds 10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back to back of standard size 75×25 mm, 75×38 mm and 75×51 mm.
Documents
Specifications
  • Description:
    Staining rack, only, for 10 to 20 slides
  • W×D×H:
    - mm