Order Entry
Belgium
Orders LinkContactUsLinkComponent
DNPH reference materials

DNPH reference materials

Formaldehyde-2,4-DNPH, 100 µg/ml, CH₃CN, 1 ml/ampoule, 1 mL
Catalog # REST33082
Supplier:   Restek
DNPH reference materials
Catalog # REST33082
Supplier:  Restek
CAS Number:  
DNPH reference materials

Formaldehyde-2,4-DNPH, 100 µg/ml, CH₃CN, 1 ml/ampoule, 1 mL

CAS Number:  
Price
64,40 €
Log in for availability
1 * 1 mL
Product Details & Documents

The reported concentrations reflect the amount of aldehyde or ketone in the mixture.The concentration of derivatised aldehyde or ketone is not reported.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    CH₃CN
  • Pk:
    1 mL
  • Description:
    Formaldehyde-2,4-DNPH
  • Pk Info:
    1 ml/ampoule

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.