Microplates, 96-well, non binding
96-well microplate, F-bottom, non binding, chimney well, solid bottom, Black
Catalog # 738-0169
Supplier: Greiner Bio-One
Microplates, 96-well, non binding
Catalog # 738-0169
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Specifications
- Colour:Black
- Description:96-well microplate, F-bottom, non binding, chimney well, solid bottom
- Seq:05
- Cat. No.:738-0169
- Environmental certification:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact