Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Forceps

VWR® Forceps

Square end, 150 mm
Catalog # 232-0152
Supplier:   VWR Collection
VWR® Forceps
Catalog # 232-0152
Supplier:  VWR Collection
VWR® Forceps
VWR® Forceps

Square end, 150 mm

Price
47,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

PTFE, white.

  • Very good chemical resistance, inert, non stick
  • Temperature resistant from –200 to +280 °C
  • Smooth surfaces prevent contamination and allow easy cleaning
Specifications
  • Type:
    Square end
  • Length:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.