Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Sample Vials

VWR® Sample Vials

25,00 ml, 33×64 mm, Yes
Catalog # 215-1900
Supplier:   VWR Collection
VWR® Sample Vials
Catalog # 215-1900
Supplier:  VWR Collection
VWR® Sample Vials
VWR® Sample Vials

25,00 ml, 33×64 mm, Yes

Price
109,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These sample vials are made from PTFE.

  • Ideal for storing aggressive or expensive materials
  • Suitable for small scale reactions at low pressure
  • Tapered inner for easy removal of contents
  • Thick wall construction
Specifications
  • Cap included:
    Yes
  • Ø×H:
    33×64 mm
  • Capacity:
    25,00 ml
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.