TURBIDITY STANDARD 100/123 NTU 1 GALLON 1 * 1 SET
Catalog # YSIA607300
Supplier: YSI ENVIRONMENTAL
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Tubing, rubber, vacuum
Supplier: VACUUBRAND