Orders LinkContactUsLinkComponent
Accessories for data loggers, Dickson®
DicksonWare™ software and USB cable, for data loggers
Catalog # 620-1515
Supplier:   DICKSON
Accessories for data loggers, Dickson®
Catalog # 620-1515
Supplier:  DICKSON
Accessories for data loggers, Dickson®
DicksonWare™ software and USB cable, for data loggers
Price
167,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DicksonWare™ software (incl. USB or serial cable) is needed for setting up Dickson loggers. They enable viewing and downloading data, and resetting the logger to the factory settings.
Specifications
  • Description:
    DicksonWare™ software and USB cable, for data loggers
  • Cat. No.:
    620-1515
  • Interface:
    USB