Flasks, flat bottom with standard ground joint
500 ml, 29/32 NS
Catalog # NEUB0752-29-0500
Supplier: NEUBERT VOLUME GLASSWAERE
Flasks, flat bottom with standard ground joint
Catalog # NEUB0752-29-0500
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Borosilicate glass 3,3, amber.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.
Specifications
- Socket size:29/32 NS
- Capacity:500 ml
- Seq:05