Orders LinkContactUsLinkComponent
Transmitted light
Transmitted light
VisiLED Transmitted light stage TLS-BF, ext.Ø 84 mm, max. 20,000 cd/m² (luminance in center)
Catalog # 630-3505
Transmitted light
Catalog # 630-3505
Transmitted light
Transmitted light
VisiLED Transmitted light stage TLS-BF, ext.Ø 84 mm, max. 20,000 cd/m² (luminance in center)
Price
607,00 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    VisiLED Transmitted light stage TLS-BF, ext.Ø 84 mm, max. 20,000 cd/m² (luminance in center)
  • Seq:
    03