Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Mortars without pestles

120 ml, 75 mm, 100 mm
Catalog # BIBB718242
Supplier:   SciLabware
Mortars without pestles
Catalog # BIBB718242
Supplier:  SciLabware
Mortars without pestles
Mortars without pestles

120 ml, 75 mm, 100 mm

Price
76,10 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Ideal for mixing and grinding powders.

  • Borosilicate glass
  • Low form model

Ordering information: Please order pestles separately.

Spécifications
  • Ø:
    100 mm
  • Capacity:
    120 ml
  • Height:
    75 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.