PCR plates, Eppendorf twin.tec®
Product Details & Documents
PP wells with PC plate deck and skirt. The rigidity of the plate makes it ideal for robotic handling and the PP wells have an inert, non-wetting surface that prevents DNA, RNA and enzymes from binding. The ultra thin walls ensure optimum heat transfer into the sample. All plates are autoclavable and certified free from DNase, RNase and human genomic DNA.
Eppendorf twin.tec® PCR Plate 96, skirted
- Compatible with automated systems, fits most thermal cyclers and the MegaBACE capillary sequencer
- Within SBS footprint requirements (127,76 mm (±0,25 mm) × 85,47 mm (±0,25 mm))
- Low profile design for enhanced efficiency of PCR and enables the highest efficiency for small sample volumes
- 150 µl maximum well volume (when used with cap strips)
Eppendorf twin.tec® PCR Plate 96, semi-skirted
- Suitable for Real-Time PCR
- Fits most thermal cyclers, including ABI GeneAmp 9700
- 250 µl maximum well volume (when used with cap strips)
Eppendorf twin.tec® PCR Plate 384
- U-shaped well for easy identification
- Continuous skirt for compatibility with automated systems
- Within SBS footprint requirements (127,76 mm (±0,25 mm)×85,47 mm (±0,25 mm))
- Suitable for all common 384-well thermal cyclers, particularly Mastercycler 384
- 45 µl maximum well volume
Specifications
- Frame colour:Orange
- No. of wells:384
- Profile:Standard
- Skirt:Fully skirted
- Well colour:Clear
- Seq:02
- Well volume:0,04 ml
- Cat. No.:713-0011
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



