Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions

Buffer standard DIN 19266, 6,865, -, 25 °C, Plastic bottle, 500 mL
  REGE10687
 :   REAGECON
Buffer solutions
  REGE10687
 :  REAGECON
 :  
Buffer solutions
Buffer solutions

Buffer standard DIN 19266, 6,865, -, 25 °C, Plastic bottle, 500 mL

 :  
 
48,70 €
   
1 * 500 mL
 

 

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • Description:
    Buffer standard DIN 19266
  • pH accuracy:
    -
  • pH value:
    6,865

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.