Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tip refill system, Axygen®

Tip refill system, Axygen®

Empty refill racks, RFL-R-1200, 1200 µl refill inserts
Catalog # 732-1403
Supplier:   Corning
Tip refill system, Axygen®
Catalog # 732-1403
Supplier:  Corning
Tip refill system, Axygen®
Tip refill system, Axygen®

Empty refill racks, RFL-R-1200, 1200 µl refill inserts

Price
52,20 €
Log in for availability
1 * 10 ST
Product Details & Documents

The Axygen® Tip Refill System contains durable refill racks that can be easily refilled by hand and are autoclavable.

  • RNase-/DNase-free
  • Non pyrogenic
  • Translucent rack allows tips to be easily seen
  • Colour-coded insert allows for easy identification of tip
Specifications
  • Description:
    Empty refill racks, RFL-R-1200
  • Seq:
    02
  • For use with:
    1200 µl refill inserts
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.