Orders LinkContactUsLinkComponent
Poly(α-methylstyrene) analytical standards, for GPC
Poly(α-methylstyrene) analytical standards, for GPC
~106000 Mn, Mw/Mn ~1,02, 250 mg
Catalog # 81520-250MG
Supplier:   Merck
Poly(α-methylstyrene) analytical standards, for GPC
Catalog # 81520-250MG
Supplier:  Merck
CAS Number:  
Poly(α-methylstyrene) analytical standards, for GPC
Poly(α-methylstyrene) analytical standards, for GPC
~106000 Mn, Mw/Mn ~1,02, 250 mg
CAS Number:  
Price
161,00 €
Log in for availability
1 * 250 mg
Product Details & Documents
Poly(α-methylstyrene) may find applications as a radical producing agent, in the thermal degradation of polystyrene.
Certifications: Exact values can be taken from the accompanying certificate of analysis.
Caution: Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.
Specifications
Specifications
  • Pk:
    250 mg
  • Description:
    ~106000 Mn, Mw/Mn ~1,02
  • Cat. No.:
    81520-250MG
  • M-Clarity quality level:
    MQ100