Poly(α-methylstyrene) analytical standards, for GPC
~7800 Mn, Mw/Mn ~1,07, 250 mg
Catalog # 81516-250MG
Supplier: Merck
Poly(α-methylstyrene) analytical standards, for GPC
Catalog # 81516-250MG
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Poly(α-methylstyrene) may find applications as a radical producing agent, in the thermal degradation of polystyrene.
Certifications: Exact values can be taken from the accompanying certificate of analysis.
Caution: Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.
Specifications
Specifications
- Pk:250 mg
- Description:~7800 Mn, Mw/Mn ~1,07
- Cat. No.:81516-250MG
- M-Clarity quality level:MQ100