Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Cyclohexane, Supelco®

Glass bottle, 5 mL
Catalog # 28920-5ML
Flammable
Health
Irritant
Environment
Supplier:   Merck
Cyclohexane, Supelco®
Catalog # 28920-5ML
Supplier:  Merck
CAS Number:  110-82-7
Cyclohexane, Supelco®
Cyclohexane, Supelco®

Glass bottle, 5 mL

CAS Number:  110-82-7
Price
25,10 €
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • UN:1145
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-806-2

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.