Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cyclohexane, Supelco®

Cyclohexane, Supelco®

Glass bottle, 10 mL
Catalog # 28920-10ML
Flammable
Health
Irritant
Environment
Supplier:   Merck
Cyclohexane, Supelco®
Catalog # 28920-10ML
Supplier:  Merck
CAS Number:  110-82-7
Cyclohexane, Supelco®
Cyclohexane, Supelco®

Glass bottle, 10 mL

CAS Number:  110-82-7
Price
42,20 €
Log in for availability
1 * 10 mL
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • UN:1145
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-806-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL

Specifications