COLOUR REFERENCE SOL. B ACC. TO PH EUR,SET (AMP. WITH 10ML) 1 * 1 KIT
Catalog # SUPL92936-1SET-F
Supplier: Merck
COLOUR REFERENCE SOL. B ACC. TO PH EUR,SET (AMP. WITH 10ML) 1 * 1 KIT
Catalog # SUPL92936-1SET-F
Supplier: Merck
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Erlenmeyer flasks, narrow or wide neck, Super Duty, DURAN®
Supplier: DWK Life Sciences
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck