Silver protein
25 g
Catalog # 05495-25G
Supplier: SIGMA ALDRICH MICROSCOPY
Jump to:
Product Details & Documents
Protargol silver impregnation and neuronal selectivity for light and electron microscopic observations.
- Assay: ~8% Ag basis
- Titration: 7,0 to 9,0%
- pH: 8,0 to 9,5 (10 mg/ml in water)
- Solubility: 1 g in 10 ml of water
Silver proteinate (protargol) is used as a component of the Periodic Acid-ThioSemiCarbazide-Silver Proteinate (PA-TSC-SP) staining protocol for ultrastructural detection of carbohydrates such as glycogen, and proteoglycans.
Synonyms: Silver colloidal, Silver nucleate, Silvol, Mild protargin, Colloidal Silver
- CAS Number:9015-51-4
- Formula:Ag (with protein)
- MDL Number:MFCD00132268
Specifications
Specifications
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



