Order Entry
Belgium
Orders LinkContactUsLinkComponent
Acetone, Supelco®

Acetone, Supelco®

Glass bottle, 5 mL
Catalog # 90872-5ML-F
Flammable
Irritant
Supplier:   Merck
Acetone, Supelco®
Catalog # 90872-5ML-F
Supplier:  Merck
CAS Number:  67-64-1
Acetone, Supelco®
Acetone, Supelco®

Glass bottle, 5 mL

CAS Number:  67-64-1
Price
169,00 €
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • UN:1090
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • REACH:01-2119471330-49

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.