Safety spectacles, A700
Anti-UV, anti-scratch, Transparent, Clear PC
Catalog # 111-0167
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
The sporty single lens A700 safety glasses provide a stylish fit for everyday application. The optional Fog-Ban minimises lens fogging even in extreme conditions.
- Lightweight design with sport temples and soft temple tip pads
- Secure, wrap-around fit and all-day comfort
- Moulded universal nose bridge
- Eight base polycarbonate hardcoated lenses
- 180° field of distortion-free vision
It is suitable for use in food industries, logistics, maintenance, telecoms, transportation, utilities, industry, building and construction, catering.
Certifications: EN 166, EN 170
Specifications
- Type:Anti-UV, anti-scratch
- Colour:Transparent
- Lenses:Clear PC