Orders LinkContactUsLinkComponent
Safety spectacles, A800
Safety spectacles, A800
Anti-Fog, Grey, Clear PC
Catalog # 111-0402
Supplier:   HONEYWELL SAFETY
Safety spectacles, A800
Catalog # 111-0402
Supplier:  HONEYWELL SAFETY
Safety spectacles, A800
Safety spectacles, A800
Anti-Fog, Grey, Clear PC
Price
5,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Single lens safety glasses for general protection against impact.
  • Temples with soft and padded inserts with extra flex built in
  • Slim profile for smaller faces
  • Sporty styling provides a fashionable fit
Suitable for applications in utilities, manufacturing, biotech, laboratories, cleanrooms, ship building and automotive manufacturing.
Certifications: EN 166.
Specifications
  • Type:
    Anti-Fog
  • Colour:
    Grey
  • Seq:
    02
  • Lenses:
    Clear PC