Orders LinkContactUsLinkComponent
Flame atomisation systems
Air/acetylene burner assembly
  VARI210164000
 :   VARIAN
Flame atomisation systems
  VARI210164000
 :  VARIAN
Flame atomisation systems
Flame atomisation systems
Air/acetylene burner assembly
1 790,00 €
1 * 1 ST
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
  • Description:
    Air/acetylene burner assembly
  • Seq:
    301
Accessories
  •  
    Description
  • VARI6910009700
    Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"
    1 * 1 ST - 21,00 €
  •  
    Description
  • VARI9910025700
    Glass impact beads
    1 * 5 ST - 192,00 €
  •  
    Description
  • VARI9910093400
    Aqueous spray chamber o-ring
    1 * 1 ST - 81,00 €
  •  
    Description
  • VARI9910093500
    Organics o-ring
    1 * 1 ST - 1 100,00 €
  •  
    Description
  • VARI9910093600
    Fluorinated mixing paddles
    1 * 1 ST - 237,00 €
  •  
    Description
  • VARI9910105800
    Integrated liquid trap (fluorinated float and magnet)
    1 * 1 ST - 401,00 €
  •  
    Description
  • VARI9910093000
    Nebuliser capillary kit
    1 * 1 ST - 722,00 €
  •  
    Description
  • VARI9910093100
    Integrated nebuliser block
    1 * 1 ST - 1 280,00 €
  •  
    Description
  • VARI190034100
    Flame atomisation system kit
    1 * 1 ST - 2 720,00 €