Flame atomisation systems
Burner cleaning and alignment card assembly
Catalog # VARI9910053900
Supplier: VARIAN
Jump to:
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
- Contoured design for outstanding resistance to blockage even with the most difficult examples
- Corrosion-resistant components
- For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Specifications
- Description:Burner cleaning and alignment card assembly
- Seq:303
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice
- VARI6910009700Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"Log in for availability1 * 1 ST - 21,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- VARI9910105800Integrated liquid trap (fluorinated float and magnet)Log in for availability1 * 1 ST - 401,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice