Orders LinkContactUsLinkComponent
Accessories for flame atomisation systems
Accessories for flame atomisation systems
Glass impact beads
Catalog # VARI9910025700
Supplier:   VARIAN
Accessories for flame atomisation systems
Catalog # VARI9910025700
Supplier:  VARIAN
Accessories for flame atomisation systems
Accessories for flame atomisation systems
Glass impact beads
Price
192,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
  • Contoured design for outstanding resistance to blockage even with the most difficult examples
  • Corrosion-resistant components
  • For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
Ordering information: Ordering information: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Specifications
  • Description:
    Glass impact beads
  • Seq:
    903