Flame atomisation systems
Cleaning wire (Lר = 3×0,3 m)
Catalog # VARI9910024700
Supplier: VARIAN
Jump to:
Product Details & Documents
The Mark 7 atomisation system handles complex matrices and achieves precise results quickly.
- Contoured design for outstanding resistance to blockage even with the most difficult examples
- Corrosion-resistant components
- For high-acid matrices
Mark 7 system includes removable twin-headed mixing paddles in order to offering thorough mixing, super-fine aerosol and for precise, accurate determinations.
This system provides the capability to introduce high matrix samples and the flexibility to achieve high sensitivity. In this system the operator can tune performances by the external adjustable impact beads.
: An adjustable barrel nebuliser, nebuliser-cleaning wire, PTFE impact beads, a burner cleaning and alignment card, and high-vacuum capillary tubings are available separately.
Specifications
- Description:Cleaning wire (Lר = 3×0,3 m)
- Seq:202
Accessories
Product Information
- Catalog #AvailabilityPrice
- Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"Nitrile O-ring for capillary assembly in spray chamber ; i.Ø = 1/32" o.Ø = 3/32"VARI6910009700Log in for availability1 * 1 ST - 21,00 €
Product Information
- Catalog #AvailabilityPrice
- Glass impact beads
Product Information
- Catalog #AvailabilityPrice
- Aqueous spray chamber o-ring
Product Information
- Catalog #AvailabilityPrice
- Organics o-ring
Product Information
- Catalog #AvailabilityPrice
- Fluorinated mixing paddles
Product Information
- Catalog #AvailabilityPrice
- Integrated liquid trap (fluorinated float and magnet)Integrated liquid trap (fluorinated float and magnet)VARI9910105800Log in for availability1 * 1 ST - 401,00 €
Product Information
- Catalog #AvailabilityPrice
- Nebuliser capillary kit
Product Information
- Catalog #AvailabilityPrice
- Integrated nebuliser block
Product Information
- Catalog #AvailabilityPrice
- Flame atomisation system kit