Orders LinkContactUsLinkComponent
Rack overlays for sample racks
Rack overlays for sample racks
Rack overlay kit for 16 mm O.D. tube rack (allows 60, 13 mm OD tubes to be used in the rack)
Catalog # VARI6610026100
Supplier:   VARIAN
Rack overlays for sample racks
Catalog # VARI6610026100
Supplier:  VARIAN
Rack overlays for sample racks
Rack overlays for sample racks
Rack overlay kit for 16 mm O.D. tube rack (allows 60, 13 mm OD tubes to be used in the rack)
Price
96,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Rack overlay kit for 16 mm O.D. tube rack (allows 60, 13 mm OD tubes to be used in the rack)
  • Seq:
    01