Order Entry
Belgium
Orders LinkContactUsLinkComponent
2-Picoline-[D₇] (98% D)

2-Picoline-[D₇] (98% D)

Glass bottle for solids, 100 mg
Catalog # PROOCILDLM12930.1
2-Picoline-[D₇] (98% D)
Catalog # PROOCILDLM12930.1
CAS Number:  93951-93-0
2-Picoline-[D₇] (98% D)

Glass bottle for solids, 100 mg

CAS Number:  93951-93-0
Price
236,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  2-Picoline-[²H₇]
  • CAS Number:93951-93-0
  • Formula:C₆D₇N
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications