Order Entry
Belgium
Orders LinkContactUsLinkComponent
Decontamination solution, RNase-ExitusPlus™

Decontamination solution, RNase-ExitusPlus™

RNase-ExitusPlus™, spray bottle, 500 mL
Catalog # A7153.0500
Supplier:   PanReac AppliChem
Decontamination solution, RNase-ExitusPlus™
Catalog # A7153.0500
Supplier:  PanReac AppliChem
CAS Number:  
Decontamination solution, RNase-ExitusPlus™
Decontamination solution, RNase-ExitusPlus™

RNase-ExitusPlus™, spray bottle, 500 mL

CAS Number:  
Price
97,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

RNase-ExitusPlus™ is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution that is highly active against RNase contamination. RNase-ExitusPlus™ has been demonstrated to inactivate more than 20 μg of RNase A dried onto the bottom of a microcentrifuge tube. RNase-ExitusPlus™ is stable for approximately 18 months and heat resistant. It should be stored at room temperature; at colder temperatures a precipitate may form which is easily brought into solution at 37 °C. RNase-ExitusPlus™ is supplied in spray bottles and a one litre refill pack is also available.

  • Catalytic and cooperative effects of the components cause a very rapid non-enzymatic, non-sequence-specific degradation of protein and RNase molecules
  • All components of RNase-ExitusPlus™ are readily and not harmful or toxic for humans
  • No aggressive mineral acids or alkaline substances are used; equipment and materials are not damaged or corroded even after prolonged incubation times
  • No organic solvents or volatile components, no toxic fumes
  • Elevated temperatures above approx. 50 °C speed up the reaction and the efficiency
Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    RNase-ExitusPlus™, spray bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.