Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digitonin

Digitonin

Glass bottle for solids, 5 g
Catalog # A1905.0005
Supplier:   PanReac AppliChem
Digitonin
Catalog # A1905.0005
Supplier:  PanReac AppliChem
CAS Number:  11024-24-1
Digitonin

Glass bottle for solids, 5 g

CAS Number:  11024-24-1
Price
1 650,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • EINECS:234-255-6
  • UN:3462
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.