Digitonin
Glass bottle for solids, 1 g
N° de catalogue A1905.0001
Fournisseur: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: Digitonin
- CAS Number:11024-24-1
- Formula:C₅₆H₉₂O₂₉
- MDL Number:MFCD00077729
- Molecular Weight:1229.312 g/mol
- EINECS:234-255-6
- UN:3462
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



