Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

Glass bottle for solids, 1 g
Catalog # ACRO229280010
Irritant
Health
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
Catalog # ACRO229280010
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

Glass bottle for solids, 1 g

CAS Number:  54827-17-7
Price
68,50 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Boiling Pt:402 °C (1013 hPa)
  • Melting Pt:167…170 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6

Documents

Specifications

Specification Test Results

  • Appearance:
    White to cream to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    167 to 172 °C
  • Loss on drying:
    ≤0.5 %
  • Sulfated ash:
    ≤0.2 %
  • HPLC:
    ≥99.0 %
  • Titration with HClO4:
    98.0 to 102.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications