Cyclohexane 99.5% for analysis
Plastic drum, 25 L
Supplier: Thermo Fisher Scientific
Cyclohexane 99.5% for analysis
Catalog # 176810250.
Supplier: Thermo Fisher Scientific
CAS Number: 110-82-7
Aller à :
Product Details & Documents
Synonymes: Hexahydrobenzene, Hexamethylene
- Numéro CAS:110-82-7
- Formule:C₆H₁₂
- Numéro MDL:MFCD00003814
- Poids moléculaire:84.16 g/mol
- Index Merck:13,02752
- Point d'ébullition:81 °C (1013 hPa)
- Point de fusion:6.5 °C
- Densité:0.77 g/cm³
- Point d'éclair:–18 °C
- Température de stockage:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Documents
ACRO17681
SDS
Spécifications
Résultats des tests de spécification
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Substance darkened by H2SO4:≤10 APHA
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Potassium (K):≤0.2 ppm
- Sodium (Na):≤1 ppm
- Zinc (Zn):≤0.1 ppm
Spécifications
- Pack type:Plastic drum
- Pk:25 L
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



