Cyclohexane 99.5% for analysis
Plastic drum, 5 L
: Hexahydrobenzene, Hexamethylene
- :110-82-7
- :C₆H₁₂
- :MFCD00003814
- :84.16 g/mol
- :13,02752
- :81 °C (1013 hPa)
- :6.5 °C
- :0.77 g/cm³
- :–18 °C
- :Ambient
- :203-806-2
- :1145
- :3,II
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Substance darkened by H2SO4:≤10 APHA
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Potassium (K):≤0.2 ppm
- Sodium (Na):≤1 ppm
- Zinc (Zn):≤0.1 ppm
- Pack type:Plastic drum
- Pk:5 L



