Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

9,2, ±0,02, Borate, Colourless, Plastic bottle, 1 L
Catalog # ACRO383870010
Buffer solutions, ready to use
Catalog # ACRO383870010
CAS Number:  
Buffer solutions, ready to use

9,2, ±0,02, Borate, Colourless, Plastic bottle, 1 L

CAS Number:  
Price
44,10 €
Log in for availability
1 * 1 L
Product Details & Documents
  • Traceable to NIST
  • Clear colourless solutions
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    1 L
  • Colour:
    Colourless
  • pH accuracy:
    ±0,02
  • pH value:
    9,2
  • Cat. No.:
    ACRO383870010
  • Density:
    1

Specifications