Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, ready to use

Buffer solutions, ready to use

4,00, ±0,02, Phthalate, Colourless, Plastic bottle, 1 L
Catalog # ACRO383830010
Buffer solutions, ready to use
Catalog # ACRO383830010
CAS Number:  
Buffer solutions, ready to use

4,00, ±0,02, Phthalate, Colourless, Plastic bottle, 1 L

CAS Number:  
Price
61,30 €
Log in for availability
1 * 1 L
Product Details & Documents
  • Traceable to NIST
  • Clear colourless solutions
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phthalate
  • Pk:
    1 L
  • Colour:
    Colourless
  • pH accuracy:
    ±0,02
  • pH value:
    4,00

Specifications