Order Entry
Belgium
Orders LinkContactUsLinkComponent
Water for analysis

Water for analysis

Plastic bottle, 1 L
Catalog # ACRO389400010
   Safer Human and Environmental Health
Water for analysis
Catalog # ACRO389400010
   Safer Human and Environmental Health
CAS Number:  7732-18-5
Water for analysis
Water for analysis

Plastic bottle, 1 L

CAS Number:  7732-18-5
Price
26,40 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (760 mmHg)
  • Melting Pt:0 °C
  • Density:1 g/cm³
  • Storage Temperature:Ambient
  • EINECS:231-791-2

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Non-volatile matter:
    ≤0.0005 %
  • Residue after ign. (sulfated):
    ≤0.0002 %
  • pH:
    5 to 8
  • Absorbance:
    ≤0.01 (water, 1 cm, 254 nm)
  • Chloride (Cl):
    ≤0.1 ppm
  • Nitrate (NO3):
    ≤0.2 ppm
  • Phosphate (PO4):
    ≤0.05 ppm
  • Sulfate (SO4):
    ≤1 ppm
  • Silicate (as SiO2):
    ≤0.01 ppm
  • Aluminium (Al):
    ≤0.02 ppm
  • Ammonium (NH4):
    ≤0.01 ppm
  • Antimony (Sb):
    ≤0.01 ppm
  • Barium (Ba):
    ≤0.01 ppm
  • Bismuth (Bi):
    ≤0.01 ppm
  • Boron (B):
    ≤0.01 ppm
  • Cadmium (Cd):
    ≤0.01 ppm
  • Calcium (Ca):
    ≤0.1 ppm
  • Chromium (Cr):
    ≤0.01 ppm
  • Cobalt (Co):
    ≤0.01 ppm
  • Copper (Cu):
    ≤0.01 ppm
  • Gold (Au):
    ≤0.01 ppm
  • Iron (Fe):
    ≤0.01 ppm
  • Lithium (Li):
    ≤0.02 ppm
  • Magnesium (Mg):
    ≤0.05 ppm
  • Manganese (Mn):
    ≤0.01 ppm
  • Mercury (Hg):
    ≤0.05 ppm
  • Molybdenum (Mo):
    ≤0.01 ppm
  • Nickel (Ni):
    ≤0.01 ppm
  • Potassium (K):
    ≤0.05 ppm
  • Selenium (Se):
    ≤0.01 ppm
  • Silver (Ag):
    ≤0.01 ppm
  • Sodium (Na):
    ≤0.1 ppm
  • Strontium (Sr):
    ≤0.05 ppm
  • Tin (Sn):
    ≤0.01 ppm
  • Titanium (Ti):
    ≤0.01 ppm
  • Vanadium (V):
    ≤0.01 ppm
  • Zinc (Zn):
    ≤0.05 ppm
  • Zirconium (Zr):
    ≤0.01 ppm
  • Heavy metals (as Pb):
    ≤0.01 ppm
  • KMnO4-reducing substance:
    Passes test
  • Conductivity:
    ≤6 µS/cm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Cat. No.:
    ACRO389400010
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.