D-(-)-Ribose 99+%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:50-69-1
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00135453
- Molecular Weight:150.13 g/mol
- Merck Index:12,08371
- Boiling Pt:331 °C (1013 hPa)
- Melting Pt:88…92 °C
- Density:0,8 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-059-4
Documents
Specifications
Specification Test Results
- Appearance:White to light yellow to light beige Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (40°C, 5 h, 20 Torr)
- Sulfated ash:≤0.1 %
- HPLC:≥99.0 % (ELSD)
- Specific optical rotation:-19.2° to -20.8° (20°C, 589 nm) (c=4, H2O)
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Glass bottle
- Pk:5 g



