Orders LinkContactUsLinkComponent
RapID™ System
RapID™ System
RapID™ ANAII panel, 20
N° de catalogue OXOIR8311002
RapID™ System
N° de catalogue OXOIR8311002
Numéro CAS:  
RapID™ System
RapID™ System
RapID™ ANAII panel, 20
Numéro CAS:  
Prix
364,00 €
Log in for availability
1 * 20 ST
Product Details & Documents
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
  • Rapid, accurate identification of over 400 medically important organisms
  • Simple, one-step inoculation
  • Four-hour aerobic incubation
  • Easy-to-read colour reactions
Documents
Specifications
Specifications
  • Pk:
    20
  • Description:
    RapID™ ANAII panel