Orders LinkContactUsLinkComponent
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
FOCUS™ Extraction Buffer VI, includes urea, thiourea, CHAPS and non detergent sulphobetaine 201, 50 mL
Catalog # 786-233
Supplier:   G-Biosciences
Extraction buffers, FOCUS™
Catalog # 786-233
Supplier:  G-Biosciences
CAS Number:  
Extraction buffers, FOCUS™
Extraction buffers, FOCUS™
FOCUS™ Extraction Buffer VI, includes urea, thiourea, CHAPS and non detergent sulphobetaine 201, 50 mL
CAS Number:  
Price
213,00 €
Log in for availability
1 * 50 mL
Product Details & Documents
FOCUS™ Extraction Buffers are experimentally optimised concentrations of critical agents, buffering and stabilising agents, including urea, thiourea, Nonidet® P-40, CHAPS, and sulphobetaines (SB). They are designed to produce optimal protein extraction and improved spot resolution for 2D gel analysis.
Specifications
Specifications
  • Pk:
    50 mL
  • Description:
    FOCUS™ Extraction Buffer VI, includes urea, thiourea, CHAPS and non detergent sulphobetaine 201
Specifications