Orders LinkContactUsLinkComponent
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl), 1 gal.
Catalog # R018
Supplier:   G-Biosciences
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Catalog # R018
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
G-Biosciences Hybridization Buffer (Hybridization Neutralizing Solution)
Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl), 1 gal.
CAS Number:  
Price
169,00 €
Log in for availability
1 * 4 L
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
  • 1 M Tris·HCl (pH 7,4)
  • 1,5 M sodium chloride
Used to neutralize membrane-bound samples and stabilize probe-target duplexes. The Tris buffer maintains physiological pH, while the high salt concentration supports nucleic acid duplex stability and reduces nonspecific binding.
Documents
Specifications
Specifications
  • Pk:
    1 gal.
  • Description:
    Hybridization neutralizing solution (1 M Tris.HCl (pH 7,4), 1,5 M NaCl)