Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Dry buffer packs, JAW™

Phosphate buffered saline packs, JAW™, 1X, 1 l/pack, 2,7 mM potassium chloride, 137 mM sodium chloride and 10 mM phosphate buffer, 7,3 - 7,5, 20
Catalog # 786-289
Supplier:   G-Biosciences
Dry buffer packs, JAW™
Catalog # 786-289
Supplier:  G-Biosciences
CAS Number:  
Dry buffer packs, JAW™

Phosphate buffered saline packs, JAW™, 1X, 1 l/pack, 2,7 mM potassium chloride, 137 mM sodium chloride and 10 mM phosphate buffer, 7,3 - 7,5, 20

CAS Number:  
Price
130,00 €
Log in for availability
1 * 20 ST
Product Details & Documents

Premixed buffer powders that simply require adding water to generate a ready-to-use buffer.

 

Specifications

Specifications

  • Pk:
    20
  • Description:
    Phosphate buffered saline packs, JAW™, 1X, 1 l/pack
  • pH value:
    7,3 - 7,5
  • Contents:
    2,7 mM potassium chloride, 137 mM sodium chloride and 10 mM phosphate buffer

Specifications