ContactUsLinkComponent
Fridge gkpv6580 gastronomic 601l height 2150 x width 700 x depth 830mm stainless steel colour circulated air cooling nickel chromium shelves profipremiumline 1 * 1 items
Catalog # 471-0413
Supplier:  Liebherr
Fridge gkpv6580 gastronomic 601l height 2150 x width 700 x depth 830mm stainless steel colour circulated air cooling nickel chromium shelves profipremiumline 1 * 1 items
Catalog # 471-0413
Supplier:  Liebherr
Frequently Bought Together