Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Acetone, BAKER, J.T.Baker®

200 L
Catalog # 8003.9200ST
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   Avantor
Acetone, BAKER, J.T.Baker®
Catalog # 8003.9200ST
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone, BAKER, J.T.Baker®
Acetone, BAKER, J.T.Baker®

200 L

CAS Number:  67-64-1
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specification Test Results

  • Acidity or Alkalinity:
    passes test
  • Appearance of solution:
    passes test
  • Identification:
    passes test
  • Matter Insoluble in Water:
    passes test
  • Reducing Substances:
    passes test
  • Relative Density:
    0.790-0.793
  • Residue on Evaporation:
    max. 50 ppm
  • Water (H2O):
    max. 3 g/l
  • Additional Specification(s)::
    -
  • Assay (by GC):
    min. 99,0 %
  • Related Substances::
    -
  • Benzene:
    max. 2 ppm (v/v)
  • Methanol (CH3OH):
    max. 0,05 % (v/v)
  • Other Impurities:
    passes test
  • 2-Propanol (CH3CHOHCH3):
    max. 0,05 % (v/v)

Specifications

  • Pk:
    200 L
  • Cat. No.:
    8003.9200ST
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications