Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate

DILUT-IT™ concentrate, 7,00, ±0,02, Phosphate, Green, 1
Catalog # 4861.
Supplier:   Avantor
J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate
Catalog # 4861.
Supplier:  Avantor
CAS Number:  
J.T.Baker® DILUT-IT™, Buffer Solutions, Colour Coded, Concentrate

DILUT-IT™ concentrate, 7,00, ±0,02, Phosphate, Green, 1

CAS Number:  
Price
49,17 €
Log in for availability
1 * 1 ST
Product Details & Documents

Volumetric concentrate, for dilution to 500 ml.

  • Traceable to NIST
Specifications

Specifications

  • pH buffer type:
    Phosphate
  • Pk:
    1
  • Colour:
    Green
  • Description:
    DILUT-IT™ concentrate
  • pH accuracy:
    ±0,02
  • pH value:
    7,00

Specifications