Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Tissue processing/embedding cassettes, Swingsette®, M517

Blue
Catalog # 720-1666
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M517
Catalog # 720-1666
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M517
Tissue processing/embedding cassettes, Swingsette®, M517

Blue

Price
159,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer.

  • Hinged lid supplied in separate packaging to make it easier to use the cassettes with commercial printers; large tab to make opening easier
  • Disposable plastic cassettes are highly resistant to histological solvents
  • Oblong opening for good fluid exchange
  • Front face inclined 45° so they can be used with certain automatic labelling systems

Packaging: Each case contains two dispenser boxes of 500 cassettes each and one dispenser box of 1000 lids.

Specifications
  • Colour:
    Blue