Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Swingsette®, M517

Tissue processing/embedding cassettes, Swingsette®, M517

Green
Catalog # 720-1664
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M517
Catalog # 720-1664
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M517
Tissue processing/embedding cassettes, Swingsette®, M517

Green

Price
159,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Acetal polymer.

  • Hinged lid supplied in separate packaging to make it easier to use the cassettes with commercial printers; large tab to make opening easier
  • Disposable plastic cassettes are highly resistant to histological solvents
  • Oblong opening for good fluid exchange
  • Front face inclined 45° so they can be used with certain automatic labelling systems

Packaging: Each case contains two dispenser boxes of 500 cassettes each and one dispenser box of 1000 lids.

Specifications
  • Colour:
    Green
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.