Tissue processing/embedding cassettes, Swingsette®, M517
Purple
Catalog # 720-1656
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M517
Catalog # 720-1656
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer.
- Hinged lid supplied in separate packaging to make it easier to use the cassettes with commercial printers; large tab to make opening easier
- Disposable plastic cassettes are highly resistant to histological solvents
- Oblong opening for good fluid exchange
- Front face inclined 45° so they can be used with certain automatic labelling systems
Packaging: Each case contains two dispenser boxes of 500 cassettes each and one dispenser box of 1000 lids.
Specifications
- Colour:Purple



