Tissue processing/embedding cassettes, Swingsette®, M515
Turquoise
Catalog # 720-1630
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M515
Catalog # 720-1630
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer.
- Integral, hinged lid, with large tab for easy opening
- Disposable plastic cassettes highly resistant to histological solvents
- Flow-through slots maximise fluid exchange
- Front face inclined 45° to enable use with leading automatic labelling systems.
Packaging: Each case contains three boxes of 500 cassettes.
Specifications
- Colour:Turquoise
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



